COMH4211B95E Combi-Blocks, Inc.

Benzoic acid, 4-bromo-3-[(2-methyl-2-propen-1-yl)oxy]-, methyl ester

  • MDL number MFCD30830158
  • Cas Number : 1147531-54-1
  • Molecular Weight : 285.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5643

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5643
Availability details for
Quantity
  • Availability
  • Add to Cart