COMH4211B95F Combi-Blocks, Inc.

3-((4,4-Diethoxybutyl)carbamoyl)benzoic acid

  • MDL number MFCD31614040
  • Cas Number : 2099046-67-8
  • Molecular Weight : 309.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5646

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5646
Availability details for
Quantity
  • Availability
  • Add to Cart