COMH4211B960 Combi-Blocks, Inc.

4-(2-Hydroxy-2-methylpropoxy)-3-methylbenzoic acid

  • MDL number MFCD30290499
  • Cas Number : 1392466-98-6
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5650

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5650
Availability details for
Quantity
  • Availability
  • Add to Cart