COMH4211B961 Combi-Blocks, Inc.

5-(2,2,2-Trifluoro-ethoxy)-isophthalic acid monomethyl ester

  • MDL number MFCD29763570
  • Cas Number : 1358967-24-4
  • Molecular Weight : 278.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5656

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5656
Availability details for
Quantity
  • Availability
  • Add to Cart