COMH4211B963 Combi-Blocks, Inc.

2-[2-(Prop-2-en-1-yloxy)ethoxy]acetic acid

  • MDL number MFCD20660469
  • Cas Number : 909570-34-9
  • Molecular Weight : 160.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5658

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5658
Availability details for
Quantity
  • Availability
  • Add to Cart