COMH4211B964 Combi-Blocks, Inc.

2-Fluoro-3-(tetrahydro-2h-pyran-4-yl)pyridine

  • MDL number MFCD20528326
  • Cas Number : 1227176-92-2
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5659

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5659
Availability details for
Quantity
  • Availability
  • Add to Cart