COMH4211B965 Combi-Blocks, Inc.

Trans 2-(4-tert-butoxycarbonylamino-cyclohexylmethyl)-butyric acid

  • MDL number MFCD31583254
  • Molecular Weight : 299.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5663

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5663
Availability details for
Quantity
  • Availability
  • Add to Cart