COMH4211B966 Combi-Blocks, Inc.

4-(4-Amino-phenyl)-2,2-dimethyl-butyric acid methyl ester

  • MDL number MFCD31613961
  • Cas Number : 2270912-89-3
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-(4-aminophenyl)-2,2-dimethylbutanoate
Inchi Code 1S/C13H19NO2/c1-13(2,12(15)16-3)9-8-10-4-6-11(14)7-5-10/h4-7H,8-9,14H2,1-3H3
Inchi Key DNTAZVZAJYJBIT-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5671

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5671
Availability details for
Quantity
  • Availability
  • Add to Cart