COMH4211B96A Combi-Blocks, Inc.

Carbamic acid, n-(2-propen-1-yloxy)-, phenylmethyl ester

  • MDL number MFCD31391239
  • Cas Number : 1174758-22-5
  • Molecular Weight : 207.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5685

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5685
Availability details for
Quantity
  • Availability
  • Add to Cart