COMH4211B96B Combi-Blocks, Inc.

2-Oxo-n-(pyrimidin-2-yl)oxazolidine-3-sulfonamide

  • MDL number MFCD31555835
  • Cas Number : 1454667-36-7
  • Molecular Weight : 244.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5686

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5686
Availability details for
Quantity
  • Availability
  • Add to Cart