COMH4211B96E Combi-Blocks, Inc.

4-Methyl-1-(2-pyrazinyl)-4-piperidinamine

  • MDL number MFCD20702036
  • Cas Number : 412355-93-2
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5695

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5695
Availability details for
Quantity
  • Availability
  • Add to Cart