COMH4211B970 Combi-Blocks, Inc.

1-(1-Bromo-2,2,2-trifluoroethyl)-2,4-dichlorobenzene

  • MDL number MFCD18355500
  • Cas Number : 1342397-22-1
  • Molecular Weight : 307.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5698

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5698
Availability details for
Quantity
  • Availability
  • Add to Cart