COMH4211B972 Combi-Blocks, Inc.

[1,1'-Biphenyl]-3-amine, 4'-fluoro-4-nitro-

  • MDL number MFCD30145682
  • Cas Number : 694533-20-5
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5703

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5703
Availability details for
Quantity
  • Availability
  • Add to Cart