COMH4211B973 Combi-Blocks, Inc.

1-(4-Bromobenzyl)-piperidine-4-carbonitrile

  • MDL number MFCD11952467
  • Cas Number : 1181664-35-6
  • Molecular Weight : 279.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5705

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5705
Availability details for
Quantity
  • Availability
  • Add to Cart