COMH4211B977 Combi-Blocks, Inc.

1-Bromo-4-(2,2-difluoro-propoxy)-2-fluoro-benzene

  • MDL number MFCD31381706
  • Cas Number : 2150821-68-2
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5715

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5715
Availability details for
Quantity
  • Availability
  • Add to Cart