COMH4211B978 Combi-Blocks, Inc.

4-(2-Chloro-5-nitrobenzyl)morpholine

  • MDL number MFCD11645969
  • Cas Number : 952490-82-3
  • Molecular Weight : 256.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5717

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5717
Availability details for
Quantity
  • Availability
  • Add to Cart