COMH4211B97A Combi-Blocks, Inc.

2-Bromo-3-(propan-2-yloxy)benzaldehyde

  • MDL number MFCD29119830
  • Cas Number : 1289086-54-9
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5720

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5720
Availability details for
Quantity
  • Availability
  • Add to Cart