COMH4211B97B Combi-Blocks, Inc.

Hexyl(2-phenylethyl)amine

  • MDL number MFCD08756402
  • Cas Number : 24997-83-9
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5721

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5721
Availability details for
Quantity
  • Availability
  • Add to Cart