COMH4211B97D Combi-Blocks, Inc.

1-Pyrrolidinecarboxylic acid, 2-[[(3-hydroxypropyl)amino]carbonyl]-, 1,1-dimethylethyl ester, (2r)-

  • MDL number MFCD31381676
  • Cas Number : 1573119-55-7
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5723

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5723
Availability details for
Quantity
  • Availability
  • Add to Cart