COMH4211B97F Combi-Blocks, Inc.

tert-Butyl 2-(3-bromophenylsulfonyl)-acetate

  • MDL number MFCD31557841
  • Cas Number : 1220422-59-2
  • Molecular Weight : 335.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5725

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5725
Availability details for
Quantity
  • Availability
  • Add to Cart