COMH4211B980 Combi-Blocks, Inc.

4-(But-2-yn-1-yloxy)-1-naphthaldehyde

  • MDL number MFCD31628737
  • Cas Number : 925441-83-4
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5729

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5729
Availability details for
Quantity
  • Availability
  • Add to Cart