COMH4211B982 Combi-Blocks, Inc.

4-(3-(Piperidin-1-yl)propoxy)benzoic acid

  • MDL number MFCD09713723
  • Cas Number : 767286-87-3
  • Molecular Weight : 263.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5734

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5734
Availability details for
Quantity
  • Availability
  • Add to Cart