COMH4211B983 Combi-Blocks, Inc.

3-Amino-n-cyclopropyl-5-ethoxybenzamide

  • MDL number MFCD24046268
  • Cas Number : 1529428-63-4
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5735

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5735
Availability details for
Quantity
  • Availability
  • Add to Cart