COMH4211B985 Combi-Blocks, Inc.

Cyclopropyl-[4-(2-methoxy-ethoxy)-benzyl]-amine

  • MDL number MFCD11629934
  • Cas Number : 1095047-84-9
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5743

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5743
Availability details for
Quantity
  • Availability
  • Add to Cart