COMH4211B986 Combi-Blocks, Inc.

5-Benzyloxy-2-ethoxy-benzoic acid

  • MDL number MFCD31618904
  • Cas Number : 164161-23-3
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5746

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5746
Availability details for
Quantity
  • Availability
  • Add to Cart