COMH4211B987 Combi-Blocks, Inc.

2-Fluoro-4-isobutoxybenzaldehyde

  • MDL number MFCD30486344
  • Cas Number : 1997067-58-9
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5748

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5748
Availability details for
Quantity
  • Availability
  • Add to Cart