COMH4211B98A Combi-Blocks, Inc.

3-Bromo-n-(cyclopropylmethyl)-4-fluoroaniline

  • MDL number MFCD27980784
  • Cas Number : 1566219-42-8
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5752

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5752
Availability details for
Quantity
  • Availability
  • Add to Cart