COMH4211B98C Combi-Blocks, Inc.

Ethyl 2-chloro-4-vinylbenzoate

  • MDL number MFCD31613792
  • Cas Number : 1269027-12-4
  • Molecular Weight : 210.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5759

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5759
Availability details for
Quantity
  • Availability
  • Add to Cart