COMH4211B98D Combi-Blocks, Inc.

5-Methyl-2-vinylpyrimidine

  • MDL number MFCD31579475
  • Cas Number : 1771735-36-4
  • Molecular Weight : 120.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5764

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5764
Availability details for
Quantity
  • Availability
  • Add to Cart