COMH4211B98E Combi-Blocks, Inc.

1-Amino-n-(2,2,2-trifluoroethyl)cyclopropanecarboxamide hydrochloride

  • MDL number MFCD31613807
  • Cas Number : 1613690-18-8
  • Molecular Weight : 218.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5765

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5765
Availability details for
Quantity
  • Availability
  • Add to Cart