COMH4211B98F Combi-Blocks, Inc.

1-(2,2-Dimethylpropyl)piperidine-4-carboxylic acid

  • MDL number MFCD20952379
  • Cas Number : 1127402-21-4
  • Molecular Weight : 199.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5766

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5766
Availability details for
Quantity
  • Availability
  • Add to Cart