COMH4211B990 Combi-Blocks, Inc.

(2-Amino-6-chloro-benzyl)-carbamic acid tert-butyl ester

  • MDL number MFCD31579548
  • Cas Number : 1431960-57-4
  • Molecular Weight : 256.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5769

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5769
Availability details for
Quantity
  • Availability
  • Add to Cart