COMH4211B991 Combi-Blocks, Inc.

Methyl 3'-formyl-4'-methoxybiphenyl-4-carboxylate

  • MDL number MFCD31583172
  • Cas Number : 408372-40-7
  • Molecular Weight : 270.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5770

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5770
Availability details for
Quantity
  • Availability
  • Add to Cart