COMH4211B993 Combi-Blocks, Inc.

1H-Pyrazole, 3,5-dimethyl-1-[(3-methylphenyl)methyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-

  • MDL number MFCD28718198
  • Cas Number : 1534377-88-2
  • Molecular Weight : 326.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5774

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5774
Availability details for
Quantity
  • Availability
  • Add to Cart