COMH4211B994 Combi-Blocks, Inc.

tert-Butyl n-[2-(2-iodoethoxy)ethyl]carbamate

  • MDL number MFCD24467264
  • Cas Number : 629626-40-0
  • Molecular Weight : 315.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5775

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5775
Availability details for
Quantity
  • Availability
  • Add to Cart