COMH4211B997 Combi-Blocks, Inc.

2-Chloro-5-nitro-4-phenylpyridine

  • MDL number MFCD28757643
  • Cas Number : 1805645-34-4
  • Molecular Weight : 234.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5780

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5780
Availability details for
Quantity
  • Availability
  • Add to Cart