COMH4211B999 Combi-Blocks, Inc.

4-([4-(Trifluoromethyl)benzyl]oxy)piperidine

  • MDL number MFCD11874753
  • Cas Number : 308386-14-3
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5783

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5783
Availability details for
Quantity
  • Availability
  • Add to Cart