COMH4211B99A Combi-Blocks, Inc.

L-Leucine, n-(1-oxobutyl)-, 2,5-dioxo-1-pyrrolidinyl ester

  • MDL number MFCD31381660
  • Cas Number : 1604040-21-2
  • Molecular Weight : 298.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5785

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5785
Availability details for
Quantity
  • Availability
  • Add to Cart