COMH4211B99F Combi-Blocks, Inc.

4-Benzyloxy-2-dimethylaminomethylene-3-oxo-butyric acid ethyl ester

  • MDL number MFCD31381650
  • Cas Number : 727382-68-5
  • Molecular Weight : 291.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5794

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5794
Availability details for
Quantity
  • Availability
  • Add to Cart