COMH4211B9A0 Combi-Blocks, Inc.

4-[2-(4-Carboxy-phenyl)-ethyl]-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31614023
  • Cas Number : 906745-32-2
  • Molecular Weight : 333.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5795

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5795
Availability details for
Quantity
  • Availability
  • Add to Cart