COMH4211B9A1 Combi-Blocks, Inc.

Benzoic acid, 4-ethenyl-, 2,5-dioxo-1-pyrrolidinyl ester

  • MDL number MFCD30290789
  • Cas Number : 97856-35-4
  • Molecular Weight : 245.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5797

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5797
Availability details for
Quantity
  • Availability
  • Add to Cart