COMH4211B9A3 Combi-Blocks, Inc.

5-Amino-n-cyclopropyl-4-fluoro-2-methyl-benzamide

  • MDL number MFCD31579295
  • Cas Number : 1855130-50-5
  • Molecular Weight : 208.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-amino-N-cyclopropyl-4-fluoro-2-methylbenzamide
Inchi Code 1S/C11H13FN2O/c1-6-4-9(12)10(13)5-8(6)11(15)14-7-2-3-7/h4-5,7H,2-3,13H2,1H3,(H,14,15)
Inchi Key MZTWWSZVXBJXJK-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5809

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5809
Availability details for
Quantity
  • Availability
  • Add to Cart