COMH4211B9A4 Combi-Blocks, Inc.

N-(5-Bromo-2-fluorobenzyl)-o-methylhydroxylamine

  • MDL number MFCD21104008
  • Cas Number : 1458303-92-8
  • Molecular Weight : 234.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5813

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5813
Availability details for
Quantity
  • Availability
  • Add to Cart