COMH4211B9A8 Combi-Blocks, Inc.

2-Bromo-4-nitro-1-phenoxybenzene

  • MDL number MFCD28129735
  • Cas Number : 1071172-96-7
  • Molecular Weight : 294.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5819

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5819
Availability details for
Quantity
  • Availability
  • Add to Cart