COMH4211B9AB Combi-Blocks, Inc.

4-Bromo-2-cyclobutoxy-1-methoxymethoxybenzene

  • MDL number MFCD31579309
  • Cas Number : 944333-95-3
  • Molecular Weight : 287.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5824

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5824
Availability details for
Quantity
  • Availability
  • Add to Cart