COMH4211B9AC Combi-Blocks, Inc.

Piperidine-1,4-dicarboxylic acid 1-tert-butyl ester 4-(1-chloro-ethyl) ester

  • MDL number MFCD18074439
  • Cas Number : 1174161-99-9
  • Molecular Weight : 291.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5826

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5826
Availability details for
Quantity
  • Availability
  • Add to Cart