COMH4211B9AE Combi-Blocks, Inc.

3-Ethynyl-pyridin-2,6-diamine

  • MDL number MFCD13175935
  • Cas Number : 936342-42-6
  • Molecular Weight : 133.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5829

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5829
Availability details for
Quantity
  • Availability
  • Add to Cart