COMH4211B9AF Combi-Blocks, Inc.

tert-Butyl[3-(2-amino-5-hydroxyphenoxy)benzyl]carbamate

  • MDL number MFCD31579478
  • Cas Number : 266368-73-4
  • Molecular Weight : 330.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5832
Availability details for
Quantity
  • Availability
  • Add to Cart