COMH4211B9B0 Combi-Blocks, Inc.

N-(3-Amino-4-bromophenyl)thiophene-3-carboxamide

  • MDL number MFCD23763285
  • Cas Number : 1513147-73-3
  • Molecular Weight : 297.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5834

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5834
Availability details for
Quantity
  • Availability
  • Add to Cart