COMH4211B9B2 Combi-Blocks, Inc.

(2R)-1-Methanesulfonyl-2-methylpiperazine

  • MDL number MFCD20664430
  • Cas Number : 1032820-95-3
  • Molecular Weight : 178.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5836

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5836
Availability details for
Quantity
  • Availability
  • Add to Cart